Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Glucose monohydrate, 99+%, extra pure

Catalog No. AC450740050
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450740050 5 kg Plastic Drum
AC450740010 1 kg Plastic Bottle
AC450740250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450740050 Supplier Thermo Scientific Chemicals Supplier No. 450740050
Only null left
Add to Cart

CAS: 14431-43-7 | C6H12O6·H2O | 198.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
Molecular Weight (g/mol) 198.17

Specifications

Ignition Residue 0.1% max
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic Drum
Quantity 5 kg
Solubility Information Solubility in water: soluble.
Formula Weight 198.17
Percent Purity 99+%
Grade Extra Pure
Chemical Name or Material D(+)-Glucose monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.