Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,1,2,2-Tetrachloroethane-d2, for NMR, 99.5+ atom % D
Click to view available options
Quantity:
10 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Refractive Index | 1.4924 |
Density | 1.6100 g/mL |
Quantity | 10 mL |
Specific Gravity | 1.61 |
Boiling Point | 146.0°C |
Linear Formula | (Cl2CD)2 |
Solubility Information | Solubility aq. soln.: insoluble. Other solubilities: soluble in ether, chloroform, benzene and alcohols |
Formula Weight | 169.86 |
Grade | NMR |
Chemical Name or Material | 1,1,2,2-Tetrachloroethane-d2 |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Fatal if inhaled. Fatal in contact with skin. Toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Remove/Take off immediately all contaminated clothing. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid release to the environment. |
EINECSNumber | 251-634-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction