Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen peroxide, 12% w/v

Catalog No. AC464570010
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464570010 1 L
AC464571000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC464570010 Supplier Thermo Scientific Chemicals Supplier No. 464570010
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 7722-84-1 | H2O2 | 34.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO

Specifications

Boiling Point 100.0°C
Density 1.0500 g/mL
Quantity 1 L
Molecular Formula H2O2
Linear Formula H2O2
Specific Gravity 1.05
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 34.01
Chemical Name or Material Hydrogen peroxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May intensify fire; oxidizer.

Causes serious eye damage.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wash face, hands and any exposed skin thoroughly after handling.

Do not eat, drink or smoke when using this product.

EINECSNumber 231-765-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.