Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents

Catalog No. BP11451 Shop All Fisherbrand Products
Change view
Click to view available options
Packaging:
Glass Bottle
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
BP1145-1 Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP1145-1
This item is not returnable. View return policy

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

Glass bottle; 1L

This solvent is used in Phenol/Chloroform extractions to remove proteins from DNA or RNA samples.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroform
Melting Point -63°C
Name Note Approx. 0.75% Ethanol as Preservative
CAS 67-66-3, 64-17-5
Absorbance 0.13 max. at 254nm, 0.28 max. at 250nm, 0.8 max. at 245nm, 1 max. at 244nm
Color Colorless
Decomposition Information 290°C
Density 1.480g/cm³
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4420 to 1.4450 (at 25°C)
Assay Percent Range ≥99 % (by GC)
Residue after Evaporation 2ppm max.
Molecular Formula CHCl3
Vapor Pressure 213mbar at 20°C
MDL Number MFCD00000826
Viscosity 0.56 mPa/s at 20°C
Water 0.02% max.
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Boiling Point 61°C
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name chloroform
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38
Percent Purity ≥99%
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.