Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate Heptahydrate, Fisher BioReagents

Catalog No. BP2131 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
BP213-1 1 kg Poly Bottle
1 options

Catalog No. BP213-1

Only null left
Add to Cart

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

Poly Bottle; 1kg

Magnesium Sulfate is suitable for use in the preparation of buffer solutions.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Sulfate Heptahydrate
Melting Point 770°C
Color White
CAS 10034-99-8
pH 5 to 8.2
Physical Form Solid
Quantity 1 kg
Assay Percent Range 98.0 to 102.0 %
Molecular Formula H14MgO11S
MDL Number MFCD00149785
Merck Index 15, 5755
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate;heptahydrate
Molecular Weight (g/mol) 246.47
PubChem CID 24843
ChEBI CHEBI:31795
Formula Weight 246.47
Percent Purity 98.0 to 102.0%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure <0.1 mmHg at 20°C
Density 1.678g/cm³
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.