Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl Sulfoxide, Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Description
Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %
DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations.
Specifications
Specifications
| Melting Point | 18.4°C |
| Absorbance | ≤0.30 Absorbance Units at 280nm |
| Color | Colorless |
| Decomposition Information | >190°C |
| Density | 1.100g/cm³ |
| Boiling Point | 189°C |
| Quantity | 1 L |
| Identification | Pass Test |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:2 Instability:0 |
| Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction