Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™

Catalog No. BP2436200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
200 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2436200 200 mL Poly Bottle
BP2436-1 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2436200
Only null left
Add to Cart
  • DNase, RNase, and protease-free premixed solution containing 10% SDS for protein extraction and DNA extraction procedures
  • Reduce preparation time by using this premade SDS solution
  • SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
  • SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3 , 7732-18-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
Color Colorless
Physical Form Liquid
Boiling Point 100°C
Density 1.01g/cm³
Concentration 9.5 to 10.5%
Quantity 200 mL
Identification Pass Test
Packaging Poly Bottle
Linear Formula CH3(CH2)10CH2OSO3Na
Merck Index 15, 8768
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Grade Electrophoresis
Chemical Name or Material Sodium Dodecyl Sulfate, 10% Solution
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.