Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis-Tris, Fisher BioReagents
GSA_VA

Catalog No. BP301100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP301-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP301-100
Only null left
Add to Cart

Commonly used buffering agent

This biological buffer has a usable pH range of 5.8 to 7.2.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym 2,2-Bis(hydroxymethyl)-2,2?,2?-nitrilotriethanol, 2, 2-Bis(hydroxymethyl)-2, 2?, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Melting Point 103°C
Color White
pH 8.8 to 9.6
Physical Form Powder or Solid
Quantity 100 g
Identification Pass Test
Assay Percent Range ≥99 %
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Packaging Poly Bottle
Vapor Pressure Negligible
Formula Weight 209.24
Percent Purity ≥99%
Chemical Name or Material Bis-Tris
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.