Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride (HPLC), Fisher Chemical™

Catalog No. D143SK4 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 

HPLC mobile phase for analytical and preparative separations.

Contains approx. 15-200 ppm preservative
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Absorbance 0.01 AU max., 0.01 AU max. at 260nm, 0.014 AU max. at 254nm, 0.10 max. at 250nm, 0.2 AU max. at 240nm, 1 AU max. at 235nm, Pass Test
Color Colorless
Boiling Point 39°C
Quantity 4 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Residue after Evaporation 2ppm max.
Assay Percent Range 99.9% min by GC
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 20mmHg at 3502°C
Water 0.02% max.
Formula Weight 84.93
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Methylene Chloride, Stabilized
Show More Show Less
missing translation for 'healthHazard2' WARNING!
missing translation for 'healthHazard3' Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.