Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™

Catalog Number D1514
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl

1LT Dichloromethane, for residue analysis, Distol™, stabilised with amylene

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
  • Contains approx. 15- 200 ppm amylene as preservative
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY
Melting Point -97°C
Absorbance 1 AU max. at 233nm, 0.12 AU max. at 240nm, 0.005 AU max. at 254nm
Color Colorless
Density 1.33g/cm³
Boiling Point 39°C
Quantity 4 L
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Refractive Index 1.42 to 1.423 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range 99.9% min by GC
Titratable Acid 0.0002mEq/g max.
Vapor Pressure 20mmHg at 3502°C
UN Number 1593
Water 0.01% max.
Formula Weight 84.93
Percent Purity ≥99.9%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Methylene Chloride
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit