Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate (Pesticide), Fisher Chemical

Catalog No. E1914 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E191-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E191-4

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -83.5°C
Color Colorless
Density 0.902g/cm³
Quantity 4 L
Flash Point −4°C (25°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 103mbar at 20°C
Linear Formula CH3COOC2H5
Viscosity 0.45 cP at 20°C
UN Number 1173
Boiling Point 75°C
Formula Weight 88.11
Grade Pesticide
Chemical Name or Material Ethyl Acetate
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.