Learn More
Specifications
Specifications
| Melting Point | -83.5°C |
| Absorbance | 1 AU max. at 255nm, 0.15 AU max. at 260nm |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Safe-Cote™ Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | 0.001% max. |
| Assay Percent Range | Assay: 99.9% min by GC |
| Titratable Acid | 0.0009mEq/g max. |
| Vapor Pressure | 103mbar at 20°C |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.