Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes (HPLC), Fisher Chemical™

Catalog No. H302N119 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
19 L
Packaging:
Glass Bottle
NOWPak™ I
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H302N-119 19 L NOWPak™ I
H302-1 1 L Wide Neck Amber Bottle
H302-4 4 L Glass Bottle
H302SK-4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. H302N-119

missing translation for 'validMainframeMsg'

HPLC mobile phase for analytical and preparative separations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Absorbance 1.00 AU max. at 195nm, 0.25 AU max. at 210nm, 0.08 AU max. at 220nm
Color Colorless
Density 0.659g/cm³
Quantity 19 L
Identification Pass Test
Packaging NOWPak™ I
Physical Form Liquid
Refractive Index 1.3740 to 1.3780 (at 25°C)
Residue after Evaporation 3ppm max.
Assay Percent Range Assay: 98.5% min by GC
Vapor Pressure 160mbar at 20°C
Viscosity 0.31 mPa/s at 20°C
UN Number 1208
Water 0.02% max.
Boiling Point 69°C
Formula Weight 86.18
Percent Purity ≥98.5%
Grade Mobile phase for HPLC applications
Chemical Name or Material Hexanes
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.