Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™

Catalog No. H3064 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 

High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3, 107-83-5, 96-14-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Absorbance 0.005 AU max. at 254nm, 0.005 AU max. at 280nm, 0.07 AU max. at 220nm, 0.25 AU max. at 210nm, 1.00 AU max. at 195nm
Color Colorless
Density 0.659g/cm³
Boiling Point 69°C
Quantity 4 L
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Refractive Index 1.3710 to 1.3750 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range 95%
Vapor Pressure 160mbar at 20°C
Viscosity 0.31 mPa/s at 20°C
UN Number 1208
Water 0.01% max.
Formula Weight 86.17
Grade Also meets ACS specifications
Chemical Name or Material n-Hexane
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.