Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™

Catalog No. H306SK4 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H306SK-4 4 L Safe-Cote™ Amber Glass 
H306-1 1 L Amber Soda Lime Glass
H306-4 4 L Amber Soda Lime Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H306SK-4
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3 , 107-83-5 , 96-14-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Absorbance 0.005 AU max. at 254nm, 0.005 AU max. at 280nm, 0.07 AU max. at 220nm, 0.25 AU max. at 210nm, 1.00 AU max. at 195nm
Color Colorless
Density 0.659g/cm³
Quantity 4 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.3710 to 1.3750 (at 25°C)
Residue after Evaporation 1ppm max.
Vapor Pressure 160mbar at 20°C
Viscosity 0.31 mPa/s at 20°C
Water 0.01% max.
UN Number 1208
Boiling Point 69°C
Formula Weight 86.17
Grade Also meets ACS specifications
Chemical Name or Material n-Hexane
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.