Learn More
Specifications
Specifications
| Melting Point | -17.8°C |
| Color | Yellow |
| Density | 4.5 (Air = 1.0) |
| Boiling Point | 151.1°C |
| Flash Point | 71°C |
| Odor | Odorless |
| Assay Percent Range | >99% |
| Packaging | Steel Drum |
| Quantity | 200 L |
| UN Number | 1223 |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Combustible liquid. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Wear personal protective equipment. Ensure adequate ventilation. Keep away from open flames, hot surfaces and sources of ignition. Do not breathe vapors or spray mist. Do not ingest. Avoid contact with skin and eyes. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. NFPA Health:1 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.