Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride (Certified), Fisher Chemical™

Catalog No. L121100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
L121-100 100 g Poly Bottle
L121-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L121-100

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382°C
Melting Point 605°C
Color White
pH 7 to 8
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥98.5 %
Formula Weight 42.39
Percent Purity ≥98.5%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1.33hPa at 547°C
Density 2.060g/cm³
Assay ≥98.5%
Chemical Name or Material Lithium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.