Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

80% Acetonitrile, 20% Water with 0.1% Formic Acid, Optima LC/MS, Fisher Chemical

Catalog No. LS122500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LS122500 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LS122500
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart
  • Mobile-phase blend
  • Reduced concentration of acetonitrile improves chromatographic performance, especially of early eluting, hydrophilic peptides.
  • Pre-mixed for ease-of-use.
  • Low metal ion content to ensure minimal metal contamination.
  • Ultra filtered to ensure low levels of particulate contamination.
  • Recommended solvent for use with Thermo Scientific EASY-nLC 1200 and Ultimate RSLCnano.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetonitrile with formic acid
CAS 7732-18-5, 64-18-6
Quantity 500 mL
Odor Aromatic
Packaging Amber Glass
Physical Form Liquid
Specific Gravity 0.7810
Grade LC-MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.