Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Oil, Light (NF/FCC), Fisher Chemical™
GSA_VA

Catalog No. O12120 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O121-20 20 L Steel Pail
O121-1 1 L Glass Bottle
O121-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O121-20
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Saybolt viscosity: 158 maximum

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Acidity Pass Test (NF)
Absorbance Pass Test (FCC)
Color White
Density 0.83g/mL
Boiling Point 260°C
Flash Point 160°C
Packaging Steel Pail
Quantity 20 L
Specific Gravity 0.818 to 0.880 (FCC/NF)
Residual Solvents Meets Requirements (NF)
Viscosity 3 CS min. (FCC), 3.0 to 34.4 mm2.S-1 (NF)
Vapor Pressure <0.1 mmHg
Grade FCC/NF
Physical Form Liquid
Chemical Name or Material Mineral Oil, Light
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:0
Flammability:1
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.