Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutaraldehyde Solution, 25% (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Specifications
Specifications
| Physical Form | Liquid |
| Melting Point | -5°C |
| Boiling Point | 100°C |
| Density | 1.060g/cm³ |
| Packaging | Glass Bottle |
| Vapor Pressure | 16.4mmHg at 20°C |
| Quantity | 4 L |
| Assay Percent Range | 24.0 to 26.5 % |
| Linear Formula | HC(O)(CH2)3CHO |
| Formula Weight | 100.12 |
| Show More |
Safety and Handling
| Hazard Category | SIGNAL WORD: DANGER! |
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction. NFPA Health: 3 Flammability: 1 Instability: 1 |
| DOTInformation | DOT Class 6.1, : Poison |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction