Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide (Pellets/NF/FCC/EP/BP/JP), Fisher Chemical

Catalog No. P258212 Shop All Fisherbrand Products
Change view
Click to view available options
:
Poly Bottle; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
P258-212 Poly Bottle; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P258-212
Only null left
Add to Cart

Specifications

pH 13.5
Color Yellow
Boiling Point 1320°C
CAS 1310-58-3
Melting Point 360°C
Molecular Formula HKO
MDL Number MFCD00003553
Density 2.04g/cm³
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 2.5 kg
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11
Percent Purity 85.0 to 100.5% (EP)
Grade BP/EP/FCC/JP/NF
Identification Pass Test (BP), Pass Test (BP/EP/FCC/JP/NF), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (NF)
Packaging Poly Bottle
Residual Solvents Meets Requirements (BP), Meets Requirements (BP/EP/NF), Meets Requirements (EP), Meets Requirements (NF)
Physical Form Solid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.