Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propylene Glycol (USP/FCC), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Description
Propylene Glycol (USP/FCC)
Specifications
Specifications
| Acidity | Pass Test (FCC/USP) |
| Melting Point | -60°C |
| pH | 6 to 8 |
| Boiling Point | 187°C |
| Ignition Residue | 0.007% max. (FCC/USP) |
| Packaging | Glass Bottle |
| Linear Formula | CH3CHOHCH2OH |
| Quantity | 4 L |
| Specific Gravity | 1.035 to 1.037 (FCC, USP) |
| Residual Solvents | Meets Requirements (USP) |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting. NFPA Health:0 Flammability:1 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction