Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical™

Catalog No. SA504 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA50-4 4 L Poly Bottle
SA50-1 1 L Poly Bottle
SA50-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SA50-4
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 0.10
Concentration 0.498 to 0.502N
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Quantity 4 L
Physical Form Liquid
UN Number 1789
Grade Certified
Concentration or Composition (by Analyte or Components) 0.5N (N/2)
Chemical Name or Material Hydrochloric Acid Solution, 0.5N (N/2)
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.