Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™ Catalog No.: SA621
  • $87.65 / Each

    Save 31%

    Reg : $126.90

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $416.65 / Case of 6 EA

    Save 31%

    Reg : $603.50

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart

Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™

Catalog No. SA621
1 L, Poly Bottle
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle

Catalog No. SA62-1

  • $87.65 / Each

    Save 31%

    Reg : $126.90

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $416.65 / Case of 6 EA

    Save 31%

    Reg : $603.50

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
Only null left
Add to Cart
Request Bulk or Custom Formats

Frequently Bought Together

This Item: Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™

$$PRICE_T1_1$$

$$PRICE_T2_1$$ / $$PRICE_UNIT_T2_1$$

Save 31%

Reg : $126.90

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Save 31%

Reg : $603.50

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

$$PROMO_BADGE_1$$

Use promo code $$SAVE_CODE_1$$ at checkout

$$CLEARANCE_BADGE_1$$

Ansell Kimtech™ Purple Nitrile™ Gloves

$$TYPE_2$$

$$PRICE_T1_2$$

$$PRICE_T2_2$$ / $$PRICE_UNIT_T2_2$$

$$PROMO_BADGE_2$$

Use promo code $$SAVE_CODE_2$$ at checkout

$$CLEARANCE_BADGE_2$$

Fisherbrand™ Transfer Pipettes

$$TYPE_3$$

$$PRICE_T1_3$$

$$PRICE_T2_3$$ / $$PRICE_UNIT_T2_3$$

Save 39%

Reg : $76.50

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Save 39%

Reg : $573.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

$$PROMO_BADGE_3$$

Use promo code $$SAVE_CODE_3$$ at checkout

$$CLEARANCE_BADGE_3$$

Total price

$193.62

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate | CAS: 7647-01-0 | ClH | 36.46 g/mol

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

TRUSTED_SUSTAINABILITY

Specifications

pH 0.10
Chemical Name or Material Hydrochloric Acid Solution, 0.01N (N/100)
CAS 7647-01-0, 7732-18-5
Concentration 0.0099 to 0.0101N
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
UN Number 1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Grade Certified
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number211504 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number232657 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number215810 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number241967 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number191741 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.