Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, pH 8.00 |
| CAS | 1310-73-2, 7778-77-0, 7732-18-5 |
| pH | 8.0 |
| Physical Form | Liquid |
| Quantity | 20 L |
| Identification | Pass Test |
| Packaging | PolyPac™ |
| MDL Number | MFCD00147253 |
| Traceability to NIST | Traceable to NIST. |
| Grade | Certified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction