Learn More
Sodium Thiosulfate Pentahydrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Shop All Fisherbrand ProductsSpecifications
Specifications
| Color | Colorless |
| Boiling Point | 100°C |
| Melting Point | 0°C |
| Concentration | 0.0995 to 0.1005N |
| Grade | Certified |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Traceability to NIST | LOT NUMBER USED POTASSIUM DICHROMATE |
| Vapor Pressure | 14mmHg |
| Quantity | 4 L |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. NFPA Health:1 Flammability:0 Instability:0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.