Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Optima™, Fisher Chemical™

Catalog No. T2914 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T291-4 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T291-4
Get Up to $1,800 in Chemicals with Purchase
View all promotions
 Restricted Item
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Absorbance 0.005 AU max. at 350nm, 0.02 AU max. at 325nm, 0.1 AU max. at 300nm, 1 AU max. at 285nm
Color Colorless
Density 0.866g/cm³
Quantity 4 L
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.4930 to 1.4950 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range ≥99.8 %
Vapor Pressure 29mbar at 20°C
Linear Formula C6H5CH3
Viscosity 0.6 mPa/s at 20°C
Water 0.02% max.
Boiling Point 111°C
Formula Weight 92.14
Percent Purity ≥99.8%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Toluene
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.