Learn More
Description
- Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
- Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
- Also meets ACS specifications
- Intended for research use only
Specifications
Specifications
| Melting Point | -95°C |
| CAS | 108-88-3 |
| Absorbance | 0.005 AU max. at 350nm, 0.02 AU max. at 325nm, 0.1 AU max. at 300nm, 1 AU max. at 285nm |
| Color | Colorless |
| Density | 0.866g/cm³ |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Safe-Cote™ Amber Glass |
| Physical Form | Liquid |
| Refractive Index | 1.4930 to 1.4950 (at 25°C) |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: DANGER! |
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects. NFPA Health:2 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.