Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene (Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. T32420 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Steel Drum
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T324-20 20 L Steel Pail
T324-1 1 L Glass Bottle
T324-4 4 L Glass Bottle
T324-200 200 L Steel Drum
T324S-4 4 L Aluminum Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. T324-20
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.866g/cm³
Quantity 20 L
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99.5 %
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPa/s at 20°C
Water <0.02%
Boiling Point 111°C
Formula Weight 92.14
Percent Purity ≥99.5%
Chemical Name or Material Toluene
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health: 2
Flammability: 3
Instability: 0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.