Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water (HPLC), Fisher Chemical™

Catalog No. W5SK1 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
19 L
Packaging:
Glass Bottle
NOWPak™ I
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W5SK-1 1 L Safe-Cote™ Amber Glass 
W5-1 1 L Wide Neck Amber Bottle
W5-4 4 L Glass Bottle
W5SK-4 4 L Safe-Cote™ Amber Glass 
W5N-119 19 L NOWPak™ I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. W5SK-1
  • $179.70
    $83.65 / Each

    Save $96.05

    53% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $857.50
    $405.65 / Case of 6 EA

    Save $451.85

    53% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

Improved filtration! HPLC mobile phase for analytical and preparative separations.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Description Improved filtration! HPLC mobile phase for analytical and preparative separations. Now filtered through a 0.2 μm filter to remove particulates.
Formula Weight 18.02
Grade Mobile phase for HPLC applications
Packaging Safe-Cote™ Amber Glass 
Residue after Evaporation 1ppm max.
Vapor Pressure 17.5mmHg at 20°C
Filtered Through 0.2-micron filtered.
Density 1000g/cm³
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.