Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water (HPLC), Fisher Chemical™

Click to view available options
Quantity:
1 L
4 L
19 L
Packaging:
Glass Bottle
NOWPak™ I
Safe-Cote™ Amber Glass
Wide Neck Amber Bottle
Description
Improved filtration! HPLC mobile phase for analytical and preparative separations.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
pH | 7 |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Description | Improved filtration! HPLC mobile phase for analytical and preparative separations. Now filtered through a 0.2 μm filter to remove particulates. |
Formula Weight | 18.02 |
Grade | Mobile phase for HPLC applications |
Packaging | Safe-Cote™ Amber Glass |
Residue after Evaporation | 1ppm max. |
Vapor Pressure | 17.5mmHg at 20°C |
Show More |
Safety and Handling

Hazard Statement | Low hazard for usual industrial or commercial handling. |
Precautionary Statement | Emergency Overview Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . . NFPA Health:0 Flammability:0 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction