Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, Optima™ LC/MS Grade, Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Description
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications
Specifications
Absorbance | ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm |
pH | 7 |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Merck Index | 15, 10237 |
Formula Weight | 18.02 |
Grade | LC-MS & UHPLC-UV |
Packaging | Wide Neck Amber Bottle |
Residue after Evaporation | ≤1ppm |
Show More |
Safety and Handling
Hazard Statement | Low hazard for usual industrial or commercial handling. |
Precautionary Statement | Emergency Overview Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. NFPA Health: 0 Flammability: 0 Instability: 0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction