Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Optima™ LC/MS Grade, Fisher Chemical™

Catalog No. W64 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
W64 4 L
W6500 500 mL
W6-1 1 L
W6212 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. W64
  • PRODUCT PROMOTION
    $401.50
    $27.39 / Each

    Save $374.11

    93% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • PRODUCT PROMOTION
    $1,276.00
    $109.56 / Case of 4 EA

    Save $1,166.44

    91% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max. | CAS: 7732-18-5 | H2O | 18.015 g/mol

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.

Note: Glass bottles have fluorinated lining.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Merck Index 15, 10237
Formula Weight 18.02
Grade LC-MS & UHPLC-UV
Packaging Glass Bottle with fluorinated lining
Residue after Evaporation ≤1ppm
Vapor Pressure 17.5mmHg at 20°C
Filtered Through 0.03 micron filtered.
Density 1.000g/cm³
Certificate of Analysis Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention.

NFPA

Health: 0
Flammability: 0
Instability: 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.