Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher Apex Accessories

Catalog No. 24075970
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
24 Combi Head
24 DW Heating Block
96 DW Head
96 DW Heating Block
96 KF Heating Block
96 PCR Head
Combi Head
Heating Block 24
Heating Block 96
Matrix Heating Block
PCR Plate Heating Block
Plate Adapter
Storage Tube Adapter
UV Lamp (Replacement)
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
24-075-970 Matrix Heating Block
24-079-910 96 PCR Head
24-079-920 Combi Head
24-079-930 96 DW Head
24-079-940 24 Combi Head
24-075-910 PCR Plate Heating Block
24-075-920 96 KF Heating Block
24-075-930 96 DW Heating Block
24-075-940 24 DW Heating Block
24-075-950 Heating Block 96
24-075-960 Heating Block 24
N21445 Storage Tube Adapter
N21446 Plate Adapter
N21447 UV Lamp (Replacement)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 24-075-970 Supplier Thermo Scientific™ Supplier No. 24075970
May include imposed supplier surcharges.
Add to Cart
Add to Cart

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
TRUSTED_SUSTAINABILITY

Specifications

Product Type Matrix Heating Block
Disposable Reusable
For Use With (Application) DNA/RNA, protein and cell purification
For Use With (Equipment) KingFisher Apex
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.