Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, FOR KETONES AND ALDEHYDES | 500 ML | GFS CHEMICALS, INC. Item #: 99813 | Bulk #: 1619
SDP
Catalog No. NC9076632
Encompass_Preferred
WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, FOR KETONES AND ALDEHYDES | Substitutes for 1612 Vessel solution when moisture is to be determined in ketones. Does not contain methanol.
Catalog No. NC9076632 Supplier GFS Chemicals Inc Supplier No. 99813
WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, FOR KETONES AND ALDEHYDES | Substitutes for 1612 Vessel solution when moisture is to be determined in ketones. Does not contain methanol.