Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, DIAPHRAGMLESS, CHLOROFORM-FREE | 500 ML | GFS CHEMICALS, INC. Item #: 99856 | Bulk #: 1671
SDP
Catalog No. NC9259681
Encompass_Preferred
WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, DIAPHRAGMLESS, CHLOROFORM-FREE | Pyridine and chlorocarbon free vessel solution for coulometers without diaphragm
Catalog No. NC9259681 Supplier GFS Chemicals Inc Supplier No. 99856
WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, DIAPHRAGMLESS, CHLOROFORM-FREE | Pyridine and chlorocarbon free vessel solution for coulometers without diaphragm