Learn More
American Radiolabeled Chemicals Inc KF N-ME-3H A 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1932A50UC
Encompass_Preferred
Ketotifen fumarate-N-methyl-3H 50uCi in ethanol, 1 mCi/ml, 80-85 Ci/mmol, M.W. 425.5, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-953