Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 2 mg/ml | 6 X 1 L | GFS CHEMICALS, INC. Item #: 99624 | Bulk #: 1601
SDP
Catalog No. NC0229210
Encompass_Preferred
WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 2 mg/ml | For volumetric analysis. Suitable for general purpose or aldehydes and ketones. ; Specifications : Suitability, Titer Strength (when bottled) (mg/ml) 1.7 - 2.4 mg/mL
Catalog No. NC0229210 Supplier GFS Chemicals Inc Supplier No. 99624
WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 2 mg/ml | For volumetric analysis. Suitable for general purpose or aldehydes and ketones. ; Specifications : Suitability, Titer Strength (when bottled) (mg/ml) 1.7 - 2.4 mg/mL