Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER SOLVENT, METHANOL-FREE | 4 L | GFS CHEMICALS, INC. Item #: 99728 | Bulk #: 1609
SDP
Catalog No. NC1625569
Encompass_Preferred
WATERMARK KARL FISCHER SOLVENT, METHANOL-FREE | For volumetric analysis. To be used with reagents 1600, 1601 and stable titrant 1602-1604. For determination of water in aldehydes and ketones in addition to general usage. ; Specifications : Sensitivity, Suitability
Catalog No. NC1625569 Supplier GFS Chemicals Inc Supplier No. 99728
WATERMARK KARL FISCHER SOLVENT, METHANOL-FREE | For volumetric analysis. To be used with reagents 1600, 1601 and stable titrant 1602-1604. For determination of water in aldehydes and ketones in addition to general usage. ; Specifications : Sensitivity, Suitability