Learn More
NEOGEN KF Streptococcus Agar, 10 kg
SDP
Catalog No. 502015371
Encompass_Preferred
KF Streptococcus Agar is used with triphenyltetrazolium chloride for the selective isolation and enumeration of fecal streptococci in a laboratory setting. KF Streptococcus Agar is not intended for use in the diagnosis of disease or other conditions in humans.KF Streptococcus Agar was developed by Kenner, Clark, and Kabler for the detection of fecal streptococci in surface waters. KF Streptococcus Agar is currently used in detecting fecal streptococci in water, foods, and other materials through the membrane filtration technique or pour plate method. Fecal streptococci are normal inhabitants in the intestines, and the presence of fecal streptococci can be used as an indication of fecal contamination. The presence of fecal streptococci is of value in determining pollution sources because certain fecal streptococci are host-specific.For more information visit https://www.neogen.com/solutions/Microbiology/kf-streptococcus-agar
Catalog No. 50-201-5371 Supplier NEOGEN Supplier No. 700003179
KF Streptococcus Agar is used with triphenyltetrazolium chloride for the selective isolation and enumeration of fecal streptococci in a laboratory setting. KF Streptococcus Agar is not intended for use in the diagnosis of disease or other conditions in humans.KF Streptococcus Agar was developed by Kenner, Clark, and Kabler for the detection of fecal streptococci in surface waters. KF Streptococcus Agar is currently used in detecting fecal streptococci in water, foods, and other materials through the membrane filtration technique or pour plate method. Fecal streptococci are normal inhabitants in the intestines, and the presence of fecal streptococci can be used as an indication of fecal contamination. The presence of fecal streptococci is of value in determining pollution sources because certain fecal streptococci are host-specific.For more information visit https://www.neogen.com/solutions/Microbiology/kf-streptococcus-agar