Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ KF Streptococcus Agar
SDP

Catalog No. 23001807 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-807 Product Type: Agar Plat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-807 Supplier Hardy Diagnostics™ Supplier No. G376
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recommended for the selective isolation and enumeration of fecal streptococci (including Enterococcus) from water and food samples by direct culture or membrane filtration

  • Includes peptone as a source of nitrogen, amino acids and carbon
  • Yeast extract provides a source of vitamins, amino acids and essential trace elements
  • Maltose and lactose provide carbon as an energy source
  • Sodium azide is a selective agent added to inhibit the growth of gram-negative microorganisms
  • Bromcresol purple dye acts as an indicator.
  • Triphenyl Tetrazolium Chloride (TTC) is a colorless redox agent added to differentiate colonies on agar or membrane filters. When irreversibly reduced to formazan by actively growing cells, TTC forms a cellular insoluble red pigment that can be exploited for more efficient enumeration

Specifications

Content And Storage 2°C to 8°C
Country of Origin United States
Additive Triphenyl Tetrazolium Chloride, Sodium Azide, Bromcresol Purple
Detects Fecal streptococci
Form Prepared Medium
Format Plate
Media Type Agar
Product Type Agar Plate
Target Organism Class Gram-positive Organisms
Test Type Water Monitoring
Fill Volume 19 mL
Formulation Maltose - 20.0gm, Proteose Peptone No. 3 - 10.0gm, Yeast Extract - 10.0gm, Sodium Glycerophosphate - 10.0gm, Sodium Chloride - 5.0gm, Lactose - 1.0gm, Sodium Azide - 0.4gm, Bromcresol Purple - 15.0mg, 1% Triphenyl Tetrazolium Chloride - 10.0ml, Agar - 20.0gm
For Use With (Application) Selective isolation and enumeration of fecal streptococci from water and food samples by direct culture or membrane filtration
Incubation Conditions 35°C
Incubation Time 46 to 48 hr.
No. per Pack 10
Packaging Type Bag
Quantity 10 Plates/Bag
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Enterococcus faecalis, Escherichia coli
Protein Source Peptone, Yeast Extract
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.