Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER WATER STANDARD, 0.05 mg/g (50 ppm) | 10 X 5 ML | GFS CHEMICALS, INC. Item #: 98981 | Bulk #: 2311
SDP
Catalog No. NC9734761
Encompass_Preferred
WATERMARK KARL FISCHER WATER STANDARD, 0.05 mg/g (50 ppm) | For checking calibration of coulometric KF equipment. Water in xylene. ; Specifications : Water (mg/g) 0.047 - 0.053 mg/g
Catalog No. NC9734761 Supplier GFS Chemicals Inc Supplier No. 98981
WATERMARK KARL FISCHER WATER STANDARD, 0.05 mg/g (50 ppm) | For checking calibration of coulometric KF equipment. Water in xylene. ; Specifications : Water (mg/g) 0.047 - 0.053 mg/g