Learn More
Across International KF25/NW25 Flange to 3/8" Hose Barb Adapter for Vacuum Connection
SDP
Catalog No. NC1505458
Encompass_Preferred
One piece stainless steel KF25/NW25 to 3/8" hose barb adapter for secure vacuum connection. Compatible with the Ai 2.5, 7.2 cfm and Welch 2.3 cfm vacuum pumps, offered by Across International.
Catalog No. NC1505458 Supplier Across International Supplier No. KF381
One piece stainless steel KF25/NW25 to 3/8" hose barb adapter for secure vacuum connection. Compatible with the Ai 2.5, 7.2 cfm and Welch 2.3 cfm vacuum pumps, offered by Across International.