Learn More
Selleck Chemical LLC KG-501 (2-naphthol-AS-E-phosphate) 10mg 18228-17-6
SDP
Supplier:  Selleck Chemical LLC S840910MG
Encompass_Preferred
KG-501 (2-naphthol-AS-E-phosphate) is a cAMP response element-binding protein (CREB) inhibitor that disrupts CREB-dependent transcription (Ki = 10 ?M) and CREB:CBP interaction (Ki = 50 ?M). It also disrupts phospho (Ser-133) CREB binding to KIX with a Ki of ?90 ?M, using concentrations of CREB that were within the linear range of the binding assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8153