Learn More
Medchemexpress LLC KG-655 | 349-58-6 | 99.9% | 230.11 | 25 G
SDP
Catalog No. 5000451219
Encompass_Preferred
KG-655 is a small molecule compound that binds to the β-folded surface and internal cavity of ARNT PAS-B, interfering with its binding to the transcriptional coactivator TACC3. It can promote the homodimerization of ARNT PAS-B and has the potential to regulate multiple ARNT-mediated signaling pathways.
  • Target: ARNT
Catalog No. 50-004-51219 Supplier Medchemexpress LLC Supplier No. 25GHYW012248
KG-655 is a small molecule compound that binds to the β-folded surface and internal cavity of ARNT PAS-B, interfering with its binding to the transcriptional coactivator TACC3. It can promote the homodimerization of ARNT PAS-B and has the potential to regulate multiple ARNT-mediated signaling pathways.
  • Target: ARNT