Learn More
ATCC KG-1; Leukemia (AML); Human (Homo sapiens)
Catalog No. 502384743
Encompass
The KG-1 cell line is made up of macrophages isolated from a bone marrow aspirate obtained from a White, 59-year-old, male with erythroleukemia that evolved into acute myelogenous leukemia. KG-1 is used in cancer, immunology, and toxicology research.
Catalog No. 50-238-4743 Supplier ATCC Supplier No. CCL246
The KG-1 cell line is made up of macrophages isolated from a bone marrow aspirate obtained from a White, 59-year-old, male with erythroleukemia that evolved into acute myelogenous leukemia. KG-1 is used in cancer, immunology, and toxicology research.