Learn More
ATCC KG-1; Myelogenous Leukemia; Human (Homo sapiens)
Catalog No. 502383483
Encompass
KG-1 is a lymphoblast-like cell that was isolated from the bone marrow of a 59-year-old, White male with acute myelogenous leukemia. The cell can be used in immune system disorders and immunology research. This cell produces high levels of human leukocyte interferon upon induction by paramyxoviridae species.
Catalog No. 50-238-3483 Supplier ATCC Supplier No. CRL8031
KG-1 is a lymphoblast-like cell that was isolated from the bone marrow of a 59-year-old, White male with acute myelogenous leukemia. The cell can be used in immune system disorders and immunology research. This cell produces high levels of human leukocyte interferon upon induction by paramyxoviridae species.