Learn More
ATCC KG-1a; Acute Myeloid Leukemia; Human (Homo sapiens)
Supplier:  ATCC CCL246.1
Encompass
KG-1a is a promyeloblast macrophage cell line that was isolated from the bone marrow of a White, 59-year-old, male patient with acute myelogenous leukemia. This cell line was deposited by DW Golde and can be used in immune system disorder research.
Catalog No. 50-238-3083