Learn More
Medchemexpress LLC KG5 | 877874-85-6 | C20H16F3N7OS | 100 MG
SDP
Catalog No. 5000372975
Encompass_Preferred
KG5 is an orally active dual PDGFRβ and B-Raf allosteric inhibitor, also inhibiting Flt3, KIT, and c-Raf. It demonstrates anticancer and antiangiogenic activities.
  • Inhibits vascular smooth muscle cells and endothelial cells viability
  • Selectively blocks S338 phosphorylation
  • Inhibits PDGFRα, PDGFRβ, Flt3, and KIT
  • Prevents tumor growth in an orthotopic renal cell carcinoma model
  • Completely blocks angiogenesis in mice
  • Disrupts angiogenesis during zebrafish embryogenesis
Catalog No. 50-003-72975 Supplier Medchemexpress LLC Supplier No. HY15198100MG
KG5 is an orally active dual PDGFRβ and B-Raf allosteric inhibitor, also inhibiting Flt3, KIT, and c-Raf. It demonstrates anticancer and antiangiogenic activities.
  • Inhibits vascular smooth muscle cells and endothelial cells viability
  • Selectively blocks S338 phosphorylation
  • Inhibits PDGFRα, PDGFRβ, Flt3, and KIT
  • Prevents tumor growth in an orthotopic renal cell carcinoma model
  • Completely blocks angiogenesis in mice
  • Disrupts angiogenesis during zebrafish embryogenesis