Learn More
Medchemexpress LLC KG5 | 877874-85-6 | C20H16F3N7OS | 50 MG
SDP
Catalog No. 5000373421
Encompass_Preferred
KG5 is an orally active dual PDGFRβ and B-Raf allosteric inhibitor, also inhibiting Flt3, KIT, and c-Raf. It demonstrates anticancer and antiangiogenic activities, preventing tumor growth and blocking angiogenesis in various models.
  • Inhibits vascular smooth muscle cells (VSMCs) and endothelial cell viability
  • Selectively blocks S338 phosphorylation
  • Inhibits PDGFRα and β, Flt3, and KIT
  • Inhibits phosphorylation of MEK and ERK in endothelial cells
  • Prevents tumor growth in an orthotopic renal cell carcinoma model
  • Completely blocks angiogenesis in mice
Catalog No. 50-003-73421 Supplier Medchemexpress LLC Supplier No. HY1519850MG
KG5 is an orally active dual PDGFRβ and B-Raf allosteric inhibitor, also inhibiting Flt3, KIT, and c-Raf. It demonstrates anticancer and antiangiogenic activities, preventing tumor growth and blocking angiogenesis in various models.
  • Inhibits vascular smooth muscle cells (VSMCs) and endothelial cell viability
  • Selectively blocks S338 phosphorylation
  • Inhibits PDGFRα and β, Flt3, and KIT
  • Inhibits phosphorylation of MEK and ERK in endothelial cells
  • Prevents tumor growth in an orthotopic renal cell carcinoma model
  • Completely blocks angiogenesis in mice